missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NOD1.
Brand: Abnova PAB13791.50ug
This item is not returnable.
View return policy
Description
Sequence: EEQGHSEMEIIPSESHPHIQLLKSNRELLVSpecifications
| NOD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NOD1. | |
| In PBS (0.02% sodium azide) | |
| CARD4/CLR7.1/NLRC1 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Recognizes Nod1. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| NOD1 | |
| NOD1 | |
| A synthetic peptide corresponding to amino acids 2-31 of human NOD1. | |
| RUO | |
| 10392 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction