Learn More
NMUR2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NMUR2.
Brand: Abnova PAB28326.100uL
Description
This gene encodes a protein from the G-protein coupled receptor 1 family. This protein is a receptor for neuromedin U, which is a neuropeptide that is widely distributed in the gut and central nervous system. This receptor plays an important role in the regulation of food intake and body weight. [provided by RefSeq
Sequence: RFQAAFQNVISSFHKQWHSQHDPQLPPAQRNIFLTECHFVELTEDIGPQFPCQSSMHNSHLPTALSSEQMSRTNYQSFHFNSpecifications
| NMUR2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NMUR2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| FM-4/FM4/NMU-R2/NMU2R/TGR-1/TGR1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 56923 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| NMUR2 | |
| NMUR2 | |
| Recombinant protein corresponding to amino acids of recombinant NMUR2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only