Learn More
NME1-NME2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NME1-NME2.
Brand: Abnova PAB27463.100uL
Description
The NME1-NME2 mRNA is a naturally occurring co-transcribed product of the neighboring NME1 and NME2 genes. The significance of this co-transcribed mRNA and the function of its predicted protein product have not yet been determined. Alternative splicing of this gene results in different transcript variants encoding distinct isoforms, but the full-length nature of each variant has not been defined. [provided by RefSeq
Sequence: MVLLSTLGIVFQGEGPPISSCDTGTMANCERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVGLKFMQSpecifications
| NME1-NME2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NME1-NME2. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| NME2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 654364 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/mL) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| NME1-NME2 | |
| NME1-NME2 | |
| Recombinant protein corresponding to amino acids of human NME1-NME2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only