Learn More
NLRP9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NLRP9.
Brand: Abnova PAB27933.100uL
Description
The protein encoded by this gene belongs to the NALP protein family. Members of the NALP protein family typically contain a NACHT domain, a NACHT-associated domain (NAD), a C-terminal leucine-rich repeat (LRR) region, and an N-terminal pyrin domain (PYD). This protein may play a regulatory role in the innate immune system as similar family members belong to the signal-induced multiprotein complex, the inflammasome, that activates the pro-inflammatory caspases, caspase-1 and caspase-5. [provided by RefSeq
Sequence: ITQLVRASVVQPQTLLTQVGIFMFGISTEEIVSMLETSFGFPLSKDLKQEITQCLESLSQCEADREAIAFQELFIGLSpecifications
| NLRP9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NLRP9. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| CLR19.1/NALP9/NOD6/PAN12 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 338321 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| NLRP9 | |
| NLRP9 | |
| Recombinant protein corresponding to amino acids of human NLRP9. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only