Learn More
NLRP8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of NLRP8.
Brand: Abnova PAB19306.100ug
Description
NALPs are cytoplasmic proteins that form a subfamily within the larger CATERPILLER protein family. Most short NALPs, such as NALP8, have an N-terminal pyrin (MEFV; MIM 608107) domain (PYD), followed by a NACHT domain, a NACHT-associated domain (NAD), and a C-terminal leucine-rich repeat (LRR) region. The long NALP, NALP1 (MIM 606636), also has a C-terminal extension containing a function to find domain (FIIND) and a caspase recruitment domain (CARD). NALPs are implicated in the activation of proinflammatory caspases (e.g., CASP1; MIM 147678) via their involvement in multiprotein complexes called inflammasomes (Tschopp et al., 2003 [PubMed 12563287]).[supplied by OMIM
Specifications
| NLRP8 | |
| Polyclonal | |
| Unconjugated | |
| Western blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| NLRP8 | |
| NLRP8 | |
| A synthetic peptide corresponding to 16 amino acids near N-terminus of human NLRP8. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of NLRP8. | |
| In PBS (0.02% sodium azide) | |
| CLR19.2/NALP8/NOD16/PAN4 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 126205 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only