Learn More
NLRP12 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of NLRP12.
Brand: Abnova PAB19309.100ug
Description
This gene encodes a member of the CATERPILLER family of cytoplasmic proteins. The encoded protein, which contains an N-terminal pyrin domain, a NACHT domain, a NACHT-associated domain, and a C-terminus leucine-rich repeat region, functions as an attenuating factor of inflammation by suppressing inflammatory responses in activated monocytes. Alternatively spliced transcript variants encoding distinct isoforms have been described but the full-length nature of some of these has not been determined. [provided by RefSeq
Specifications
| NLRP12 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| NLRP12 | |
| NLRP12 | |
| A synthetic peptide corresponding to 18 amino acids near N-terminus of human NLRP12. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of NLRP12. | |
| In PBS (0.02% sodium azide) | |
| CLR19.3/FCAS2/NALP12/PAN6/PYPAF7/RNO/RNO2 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 91662 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only