missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NLN.
Brand: Abnova PAB22595.100uL
This item is not returnable.
View return policy
Description
Neurolysin (EC 3.4.24.16) is an enzyme involved in the metabolic inactivation of bioactive peptides (Garrido et al., 1999 [PubMed 10235115]).[supplied by OMIM
Sequence: LRMTLGREVMSPLQAMSSYTVAGRNVLRWDLSPEQIKTRTEELIVQTKQVYDAVGMLGIEEVTYENCLSpecifications
| NLN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NLN. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| AGTBP/DKFZp564F123/KIAA1226 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57486 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| NLN | |
| NLN | |
| Recombinant protein corresponding to amino acids of human NLN. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction