Learn More
NLGN4X, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NLGN4X
Brand: Abnova PAB28472.100uL
Description
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. The encoded protein interacts with discs, large (Drosophila) homolog 4 (DLG4). Mutations in this gene have been associated with autism and Asperger syndrome. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq
Sequence: QYPVVNTNYGKIRGLRTPLPNEILGPVEQYLGVPYASPPTGERRFQPPEPPSSWTGIRNTTQFAAVCPQHLDERSLLHDMLPIWFTANLDTLMTYVQDQNEDCLYLNIYVPTEDDIHDQNSKKPVMVYIHGGSYMEGTGNMIDGSSpecifications
| NLGN4X | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NLGN4X | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| ASPGX2/AUTSX2/HLNX/HNLX/KIAA1260/MGC22376/NLGN/NLGN4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57502 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry(1:50-1:200) The optimal working dilution should be determined by the end user. | |
| NLGN4X | |
| NLGN4X | |
| Recombinant protein corresponding to amino acids of human NLGN4X | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only