Learn More
NKX2-2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NKX2-2.
Brand:  Abnova PAB25798.100ug
Product Code. 16045977
  • £535.00 / 100µg
Description
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor. [provided by RefSeq
Specifications
Specifications
NKX2-2
Polyclonal
Unconjugated
Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
NKX2-2
NKX2-2
A synthetic peptide corresponding to 19 amino acids at internal region of human NKX2-2.
100 μg
Primary
NKX2-2 antibody is predicted to not cross-react with other NK2 homeobox family members.
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of NKX2-2.
In PBS (0.02% sodium azide)
NKX2.2/NKX2B
Rabbit
Peptide affinity purification
RUO
4821
Human, Mouse, Rat