missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2-1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NKX2-1.
Brand: Abnova PAB25797.100ug
This item is not returnable.
View return policy
Specifications
| NKX2-1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NKX2-1. | |
| In PBS (0.02% sodium azide) | |
| BCH/BHC/NK-2/NKX2.1/NKX2A/TEBP/TITF1/TTF-1/TTF1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 7080 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunofluorescence (20 ug/mL) Immunohistochemistry (2.5 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| NKX2-1 | |
| NKX2-1 | |
| A synthetic peptide corresponding to 16 amino acids at N-terminus of human NKX2-1. | |
| 100 μg | |
| Primary | |
| NKX2-1 antibody is predicted to not cross-react with other NK2 homeobox family members. At least three isoforms of NKX2-1 are known to exist; this antibody will detect all three. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction