Learn More
NKAIN4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NKAIN4.
Brand: Abnova PAB22361.100uL
Description
NKAIN4 is a member of a family of mammalian proteins (see NKAIN1; MIM 612871) with similarity to Drosophila Nkain and interacts with the beta subunit of Na,K-ATPase (ATP1B1; MIM 182330) (Gorokhova et al., 2007 [PubMed 17606467]).[supplied by OMIM
Sequence: LKDSELLTFSLSRHRSWWRERWPGCLHEEVPAVGLGAPHGQALVSGAGCALEPSYVEALHSCLQILIALLGFVCGCQVVSVFTEEEDSFDFIGGFDPFPLYHVNEKPSSLLSKQVYSpecifications
| NKAIN4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NKAIN4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| C20orf58/FAM77A/bA261N11.2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 128414 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| NKAIN4 | |
| NKAIN4 | |
| Recombinant protein corresponding to amino acids of human NKAIN4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only