Learn More
NIPA1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NIPA1.
Brand: Abnova PAB21503.100uL
Description
This gene encodes a magnesium transporter that associates with early endosomes and the cell surface in a variety of neuronal and epithelial cells. This protein may play a role in nervous system development and maintenance. Multiple transcript variants encoding different isoforms have been found for this gene. Mutations in this gene have been associated with autosomal dominant spastic paraplegia 6. [provided by RefSeq
Sequence: HGPTNIMVYISICSLLGSFTVPSTKGIGLAAQDILHNNPSSQRASpecifications
| NIPA1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NIPA1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FSP3/MGC102724/MGC35570/SPG6 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 123606 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| NIPA1 | |
| NIPA1 | |
| Recombinant protein corresponding to amino acids of human NIPA1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only