Learn More
NGDN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NGDN.
Brand: Abnova PAB20023.100uL
Description
Neuroguidin is an EIF4E (MIM 133440)-binding protein that interacts with CPEB (MIM 607342) and functions as a translational regulatory protein during development of the vertebrate nervous system (Jung et al., 2006 [PubMed 16705177]).[supplied by OMIM
Sequence: LMYLMDLTHLILDKASGGSLQGHDAVLRLVEIRTVLEKLRPLDQKLKYQIDKLIKTAVTGSLSENDPLRFKPHPSNMMSKLSSEDEEEDEAEDDQSEASGKKSVKGVSKKYVPPRLVPVHYDETEAEREKKRLERAKRRALSSSVIRELSpecifications
| NGDN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NGDN. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| C14orf120/DKFZp564O092/LCP5/NGD/lpd-2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 25983 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| NGDN | |
| NGDN | |
| Recombinant protein corresponding to amino acids of human NGDN. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only