Learn More
NEUROD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NEUROD1.
Brand: Abnova PAB3857.400uL
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus. [provided by RefSeq
Specifications
NEUROD1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NEUROD1. | |
In PBS (0.09% sodium azide) | |
BETA2/BHF-1/NEUROD/bHLHa3 | |
Rabbit | |
Protein G purification | |
RUO | |
4760 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
NEUROD1 | |
NEUROD1 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human NEUROD1. | |
400 μL | |
Primary | |
Human | |
Liquid |
For Research Use Only