missing translation for 'onlineSavingsMsg'
Learn More

NEURL Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant NEURL.

Brand:  Abnova PAB24309.100uL

Product Code. 16094687

  • £426.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: PDLVSQSGFWAKALPEEFANEGNIIAFWVDKKGRVFHRINDSAVMLFFSGVRTADPLWALVD
Specifications

Specifications

NEURL
Polyclonal
Rabbit polyclonal antibody raised against recombinant NEURL.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
NEURL1/RNF67/h-neu/neu-1
Rabbit
Antigen affinity purification
RUO
9148
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
NEURL
NEURL
Recombinant protein corresponding to amino acids of human NEURL.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only