missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant NDUFV2.
Brand: Abnova PAB19133.100uL
This item is not returnable.
View return policy
Specifications
| NDUFV2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant NDUFV2. | |
| In serum | |
| NDUFV2 | |
| Recombinant protein corresponding to full length human NDUFV2. | |
| RUO | |
| 4729 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western blot (1:500) The optimal working dilution should be determined by the end user. | |
| NDUFV2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| It can expression in BGC823 whole cell lysate. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction