Learn More
NDUFAF3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NDUFAF3.
Brand: Abnova PAB22870.100uL
Description
This gene encodes a mitochondrial complex I assembly protein that interacts with complex I subunits. Mutations in this gene cause mitochondrial complex I deficiency, a fatal neonatal disorder of the oxidative phosphorylation system. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: LLEPRIEIVVVGTGDRTERLQSQVLQAMRQRGIAVEVQDTPNACATFNFLCHEGRVTGAALIPPPGGTSLTSLGQAAQSpecifications
| NDUFAF3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NDUFAF3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| 2P1/DKFZp564J0123/E3-3/MGC10527 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 25915 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| NDUFAF3 | |
| NDUFAF3 | |
| Recombinant protein corresponding to amino acids of human NDUFAF3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only