Learn More
NCF2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NCF2.
Brand: Abnova PAB28551.100uL
Description
This gene encodes neutrophil cytosolic factor 2, the 67-kilodalton cytosolic subunit of the multi-protein NADPH oxidase complex found in neutrophils. This oxidase produces a burst of superoxide which is delivered to the lumen of the neutrophil phagosome. Mutations in this gene, as well as in other NADPH oxidase subunits, can result in chronic granulomatous disease, a disease that causes recurrent infections by catalase-positive organisms. Alternative splicing results in two transcript variants that encode the same protein. [provided by RefSeq
Sequence: QEESSPQSDIPAPPSSKAPGRPQLSPGQKQKEEPKEVKLSVPMPYTLKVHYKYTVVMKTQPGLPYSQVRDMVSKKLELRLEHTKLSYRPRDSNELVPLSEDSMKDAWGQVKNYCLTLWCENTVGDQGFPDEPKESEKADANNQTTEPQSpecifications
| NCF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NCF2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| FLJ93058/NCF-2/NOXA2/P67-PHOX/P67PHOX | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4688 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:200-1:500) Western Blot(1:100-1:250) The optimal working dilution should be determined by the end user. | |
| NCF2 | |
| NCF2 | |
| Recombinant protein corresponding to amino acids of recombinant NCF2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only