Learn More
NCAPH, subunit H, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NCAPH, subunit H.
Brand: Abnova PAB28677.100uL
Description
This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization. [provided by RefSeq
Sequence: LHCQDYRSELLFPSDVQTLSTGEPLELPELGCVEMTDLKAPLQQCAEDRQICPSLAGFQFTQWDSETHNESVSALVDKFKKNDQVFDINAEVDESDCGDFPDGSLGDDFDANDEPDHTSpecifications
| NCAPH, subunit H | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NCAPH, subunit H. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| BRRN1/CAP-H/HCAP-H | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23397 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. | |
| NCAPH | |
| NCAPH | |
| Recombinant protein corresponding to amino acids of human NCAPH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only