missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myt1l, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Myt1l.
Brand: Abnova PAB15836.50ug
This item is not returnable.
View return policy
Description
Sequence: RATSAMKKAKLSGEQMLTIKQRASNGIENDEEIKQLDEEIKELNESNSQMEADMIKLRTQITTMESNLKTIEEENKVIEQQNESLLHELANLSQSLIHSLANIQLPHMDPINEQNFDAYVTTLTEMYTNQDRYQSPENKALLENIKQAVRGIQVSpecifications
| Myt1l | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Myt1l. | |
| In PBS (50% glycerol, 0.02% sodium azide) | |
| 2900046C06Rik/2900093J19Rik/C630034G21Rik/Nztf1/Pmng1/Png-1/mKIAA1106 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Specific to recombinant protein GX0194. This antibody detects mMYT1L protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Myt1l | |
| Myt1l | |
| Recombinant GST fusion protein corresponding to 154 amino acids of mouse Myt1l. | |
| RUO | |
| 17933 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction