missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYT1L, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MYT1L.
Brand: Abnova PAB25680.100ug
This item is not returnable.
View return policy
Specifications
| MYT1L | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MYT1L | |
| MYT1L | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human MYT1L. | |
| 100 μg | |
| Primary | |
| At least three isoforms are known to exist. This antibody will only recognize the two longest isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MYT1L. | |
| In PBS (0.05% sodium azide) | |
| NZF1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 23040 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction