Learn More
MYH7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MYH7.
Brand: Abnova PAB28402.100uL
Description
Muscle myosin is a hexameric protein containing 2 heavy chain subunits, 2 alkali light chain subunits, and 2 regulatory light chain subunits. This gene encodes the beta (or slow) heavy chain subunit of cardiac myosin. It is expressed predominantly in normal human ventricle. It is also expressed in skeletal muscle tissues rich in slow-twitch type I muscle fibers. Changes in the relative abundance of this protein and the alpha (or fast) heavy subunit of cardiac myosin correlate with the contractile velocity of cardiac muscle. Its expression is also altered during thyroid hormone depletion and hemodynamic overloading. Mutations in this gene are associated with familial hypertrophic cardiomyopathy, myosin storage myopathy, dilated cardiomyopathy, and Laing early-onset distal myopathy. [provided by RefSeq
Sequence: ALRKKHADSVAELGEQIDNLQRVKQKLEKEKSEFKLELDDVTSNMEQIIKAKANLEKMCRTLEDQMNEHRSKAEETQRSVNDLTSQRAKLQTENGELSRQLDEKEALISQLTRGKLTYTQQLEDLKRQLEEEVKAKNALAHALQSSpecifications
| MYH7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MYH7. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| CMD1S/CMH1/DKFZp451F047/MGC138376/MGC138378/MPD1/MYHCB/SPMD/SPMM | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4625 | |
| Store at 4 °C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:1000) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| MYH7 | |
| MYH7 | |
| Recombinant protein corresponding to human MYH7. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only