Learn More
MYC (phospho S62), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MYC.
Brand: Abnova PAB25882.100ug
Description
The protein encoded by this gene is a multifunctional, nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. It functions as a transcription factor that regulates transcription of specific target genes. Mutations, overexpression, rearrangement and translocation of this gene have been associated with a variety of hematopoietic tumors, leukemias and lymphomas, including Burkitt lymphoma. There is evidence to show that alternative translation initiations from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site result in the production of two isoforms with distinct N-termini. The synthesis of non-AUG initiated protein is suppressed in Burkitt's lymphomas, suggesting its importance in the normal function of this gene. [provided by RefSeq]
Sequence: P-L-Sp-P-SSpecifications
| MYC | |
| Polyclonal | |
| Unconjugated | |
| Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user. | |
| MYC | |
| MYC | |
| Synthetic phosphopeptide corresponding to residues surrounding S62 of human MYC. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of MYC. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| bHLHe39/c-Myc | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4609 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only