missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MYBPC1.
Brand: Abnova PAB25710.100ug
This item is not returnable.
View return policy
Specifications
| MYBPC1 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| MYBPC1 | |
| MYBPC1 | |
| A synthetic peptide corresponding to 12 amino acids at C-terminus of human MYBPC1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MYBPC1. | |
| In PBS (0.05% sodium azide) | |
| MYBPCC/MYBPCS/slow-type | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 4604 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction