Learn More
MVP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MVP.
Brand: Abnova PAB28550.100uL
Description
This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts. [provided by RefSeq
Sequence: VFEEVLDLVDAVILTEKTALHLRARRNFRDFRGVSRRTGEEWLVTVQDTEAHVPDVHEEVLGVVPITTLGPHNYCVILDPVGPDGKNQLGQKRVVKGEKSFFLQPGEQLEQGIQDVYVLSSpecifications
| MVP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MVP. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| LRP/VAULT1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9961 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:200-1:500) Western Blot(1:250-1:500) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| MVP | |
| MVP | |
| Recombinant protein corresponding to amino acids of recombinant MVP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only