Learn More
MTO1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MTO1.
Brand: Abnova PAB22421.100uL
Description
This gene encodes a mitochondrial protein thought to be involved in mitochondrial tRNA modification. The encoded protein may also play a role in the expression of the nonsyndromic and aminoglycoside-induced deafness phenotypes associated with a specific mutation in the mitochondrial 12S rRNA gene. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: RPDNADSRLTLRGYKDAGCVSQQRYERACWMKSSLEEGISVLKSIEFLSSKWKKLIPEASISTSRSLPVRALDVLKYESpecifications
| MTO1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MTO1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CGI-02 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 25821 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| MTO1 | |
| MTO1 | |
| Recombinant protein corresponding to amino acids of human MTO1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only