Learn More
MRPL55, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MRPL55.
Brand: Abnova PAB22040.100uL
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Multiple transcript variants encoding two different isoforms were identified through sequence analysis. [provided by RefSeq
Sequence: PVLLVKQDGSTIHIRYREPRRMLAMPIDLDTLSPEERRARLRKREAQLQSRKEYEQELSDDLHVERYRQFWTRTKKSpecifications
| MRPL55 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MRPL55. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| AAVG5835/DKFZp686D1387/L55nt/MGC61802/MRP-L55/PRO19675 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 128308 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| MRPL55 | |
| MRPL55 | |
| Recombinant protein corresponding to amino acids of human MRPL55. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only