Learn More
MRPL37 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MRPL37.
Brand: Abnova PAB21787.100uL
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. [provided by RefSeq
Sequence: SATWNRESLLLQVRGSGGARLSTKDPLPTIASREEIEATKNHVLETFYPISPIIDLHECNIYDVKSpecifications
| MRPL37 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MRPL37. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| L37mt/MGC878/MRP-L2/MRP-L37/MRPL2/RPML2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51253 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| MRPL37 | |
| MRPL37 | |
| Recombinant protein corresponding to amino acids of human MRPL37. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only