Learn More
MPDU1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MPDU1.
Brand: Abnova PAB20774.100uL
Description
This gene encodes an endoplasmic reticulum membrane protein that is required for utilization of the mannose donor mannose-P-dolichol in the synthesis of lipid-linked oligosaccharides and glycosylphosphatidylinositols. Mutations in this gene result in congenital disorder of glycosylation type If. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: AEADGPLKRLLVPILLPEKCYDQLFVQWDLLHVPCLKILLSKSpecifications
| MPDU1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MPDU1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CDGIF/FLJ14836/HBEBP2BPA/Lec35/My008/PP3958/PQLC5/SL15 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9526 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| MPDU1 | |
| MPDU1 | |
| Recombinant protein corresponding to amino acids of human MPDU1. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only