Learn More
Rabbit anti-Mouse Kappa Light Chain Secondary Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-72791PE
Product Code. 30229937
  • £319.00 / 0.50mL
Description
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kappa Light Chain
Polyclonal
PBS
Rabbit
Multi-step
RUO
Secondary
Mouse
IgG
Western Blot, ELISA, Immunohistochemistry
PE
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Mouse kappa light chain
0.5 mL
Adaptive Immunity, Immunology
3514
Store at 4°C in the dark.