missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgM Heavy Chain Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB7499
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Mouse IgM | |
| 1 mL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this reacts specifically with mouse IgM. This may cross react with IgM from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:30000, ELISA 1:1000 - 1:30000; for coating plates 1:100 - 1:500 Immunohistochemistry 1:200- 1:2000, Immunocytochemistry/Immunofluorescence 1:200- 1:2000, Immunohistochemistry-Paraffin 1:200-1:2000 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3507 | |
| Mouse | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction