missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgM Fc Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69734
216.75 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Mouse samples. It is validated for ELISA, Immunoprecipitation, Immunoelectrophoresis
Specifications
| IgM Fc | |
| Polyclonal | |
| Unconjugated | |
| Whole antiserum | |
| Highly purified homogenous IgM isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA, Immunoprecipitation, Immunoelectrophoresis | |
| LYOPH | |
| ELISA, Immunoprecipitation 1:10 - 1:500, Immunoelectrophoresis | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Secondary | |
| Mouse | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction