missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG3 Heavy Chain Secondary Antibody, Rhodamine, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73632
This item is not returnable.
View return policy
Description
IgG3 Heavy Chain Polyclonal antibody specifically detects IgG3 Heavy Chain in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| IgG3 Heavy Chain | |
| Polyclonal | |
| Rhodamine | |
| DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3502 | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| IGHG3 | |
| Mouse IgG3 heavy chain | |
| 1 mg | |
| Secondary | |
| Specificity was confirmed by ELISA at less than 1% cross reactivity against other mouse heavy or light chain isotypes. | |
| Mouse | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction