missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG1 Secondary Antibody (20) [PerCP], Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43768PCP

Product Code. 30281483

  • £392.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG1
Monoclonal
PerCP
PBS
Rabbit
Protein A purified
RUO
Mouse
Purified
ELISA
20
ELISA
IgG1, Igh-4, RGD1359539, VH7183
Recombinant Mouse IgG1 Fc Protein
0.1 mL
Secondary
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.