missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG1 Heavy Chain Secondary Antibody, Texas Red, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73704
This item is not returnable.
View return policy
Description
IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| IgG1 Heavy Chain | |
| Polyclonal | |
| Texas Red | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Specificity was confirmed by ELISA at less than 1% cross reactivity against other mouse heavy or light chain isotypes. | |
| Mouse | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Mouse IgG1 heavy chain | |
| 1 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction