Learn More
Rabbit anti-Mouse IgG (H+L), Superclonal™ Recombinant Secondary Antibody, Biotin, Invitrogen™
Rabbit Recombinant Polyclonal Secondary Antibody
Brand: Invitrogen A27024
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with goat, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Mouse IgG (H+L) Recombinant Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for ELISA, Western Blot.Specifications
Mouse IgG (H+L) | |
Recombinant Polyclonal | |
Biotin | |
Rabbit | |
Gravity column chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Recombinant full-length protein | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |