Learn More
Invitrogen™ Rabbit anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor™ 647
Description
The sensitivity and specificity of each lot is confirmed using Immunocytochemistry. Minimal cross-reactivity with goat, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Superclonal |
| Concentration | 1 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 5mM sodium azide; pH 7.2 |
| Host Species | Rabbit |
| Immunogen | Recombinant full-length protein. |
| Purification Method | Gravity column chromatography |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.