missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB720-B

232.90 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18216205

  • £291.00 / 0.50mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG (H+L)
Polyclonal
Biotin
Rabbit
Affinity Purified
RUO
Secondary
Mouse
IgG
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
2.0 mg/mL
Western Blot 1:20000 - 1:10000, ELISA 1:300000, Immunohistochemistry 1:1000 - 1:5000
Mouse IgG whole molecule
0.5 mg
Adaptive Immunity, Immunology
Secondary Antibody was prepared from monospecific antiserum by immunoaffinity chromatography using Mouse IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Rabbit Serum, Mouse IgG and Mouse Serum.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only