missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG Fc Secondary Antibody, Janelia Fluor 549, Novus Biologicals™

Product Code. 18416388 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416388 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416388 Supplier Novus Biologicals Supplier No. NBP260665JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG Fc Polyclonal antibody specifically detects IgG Fc in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fc
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Rabbit anti-Mouse IgG Fc fragment Secondary Antibody was produced by repeated immunization with mouse IgG F(c) fragment in rabbit.
Purification Method Immunogen affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.