missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG F(ab')2 Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72760
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| Unconjugated | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Mouse IgG F(ab')2 fragment | |
| 2 mg | |
| Secondary | |
| Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| 1.5 mg/mL | |
| Western Blot 1:2000 - 1:10000, ELISA 1:75000, Immunohistochemistry 1:1000 - 1:5000, Dot Blot | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against Mouse IgG F(c). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction