Learn More
MOS, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MOS.
Brand: Abnova PAB3278.400uL
Description
MOS is a serine/threonine kinase that activates the MAP kinase cascade through direct phosphorylation of the MAP kinase activator MEK (MAP2K1; MIM 176872) (Prasad et al., 2008 [PubMed 18246541]).[supplied by OMIM
Specifications
| MOS | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MOS. | |
| In PBS (0.09% sodium azide) | |
| MGC119962/MGC119963/MSV | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 4342 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| MOS | |
| MOS | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MOS. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only