Learn More
MMP13 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MMP13.
Brand: Abnova PAB16221.50ug
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The protein encoded by this gene cleaves type II collagen more efficiently than types I and III. It may be involved in articular cartilage turnover and cartilage pathophysiology associated with osteoarthritis. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. [provided by RefSeq
Specifications
| MMP13 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MMP13. | |
| In PBS (0.09% sodium azide) | |
| CLG3 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 4322 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (17 ug/mL) The optimal working dilution should be determined by the end user. | |
| MMP13 | |
| MMP13 | |
| A synthetic peptide (conjugated with KLH) corresponding to human MMP13. | |
| 50 μg | |
| Primary | |
| N-terminal domain of human . | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only