Learn More
MMADHC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MMADHC.
Brand: Abnova PAB23126.100uL
Description
This gene encodes a mitochondrial protein that is involved in an early step of vitamin B12 metabolism. Vitamin B12 (cobalamin) is essential for normal development and survival in humans. Mutations in this gene cause methylmalonic aciduria and homocystinuria type cblD (MMADHC), a disorder of cobalamin metabolism that is characterized by decreased levels of the coenzymes adenosylcobalamin and methylcobalamin. Pseudogenes have been identified on chromosomes 11 and X
Sequence: EVLLEKFINGAKEICYALRAEGYWADFIDPSSGLAFFGPYTNNTLFETDERYRHLGFSVDDLGCCKVIRHSLWGTHVVVGSIFTNATSpecifications
| MMADHC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MMADHC. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| C2orf25/CL25022 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 27249 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| MMADHC | |
| MMADHC | |
| Recombinant protein corresponding to amino acids of human MMADHC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only