Learn More
MLF1IP Rabbit anti-Bovine, Chicken, Chimpanzee, Dog, Horse, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant MLF1IP.
Brand: Abnova PAB10267.100ug
Description
The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). MLF1IP, or CENPU, is an additional factor required for centromere assembly (Foltz et al., 2006 [PubMed 16622419]).[supplied by OMIM
Specifications
| MLF1IP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant MLF1IP. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| CENP-50/CENP-U/CENP-U(50)/CENP50/CENPU/FLJ23468/KLIP1/PBIP1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This affinity purified antibody is directed against human MLF1IP protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1:650000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| MLF1IP | |
| MLF1IP | |
| Recombinant protein corresponding to C-terminus 418 amino acids of human MLF1IP. | |
| RUO | |
| 79682 | |
| Bovine, Canine, Chicken, Chimpanzee, Equine, Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only