Learn More
MKS1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MKS1.
Brand: Abnova PAB21454.100uL
Description
The protein encoded by this gene localizes to the basal body and is required for formation of the primary cilium in ciliated epithelial cells. Mutations in this gene result in Meckel syndrome type 1 and in Bardet-Biedl syndrome type 13. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: TCTTKSLAMDKVAHFSYPFTFEAFFLHEDESSDALPEWPVLYCEVLSLDFWQRYRVEGYGAVVLPATPGSHTLTVSTWRPVELGTVASpecifications
| MKS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MKS1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| BBS13/FLJ20345/MES/MKS | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54903 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| MKS1 | |
| MKS1 | |
| Recombinant protein corresponding to amino acids of human MKS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only