missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKI67 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Mki67.
Brand: Abnova PAB12128.100uL
This item is not returnable.
View return policy
Specifications
| MKI67 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Mki67. | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
| D630048A14Rik/Ki-67/Ki67 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user. | |
| Mki67 | |
| Mki67 | |
| A synthetic peptide corresponding to amino acids 1700-1800 of mouse Mki67. | |
| RUO | |
| 17345 | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction