missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIG7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MIG7.
Brand: Abnova PAB8969.100ug
This item is not returnable.
View return policy
Specifications
| MIG7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MIG7. | |
| In buffer containing 0.02% sodium azide | |
| MIG7 | |
| A synthetic peptide corresponding to N-terminus human MIG7. | |
| RUO | |
| 723788 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:10000) Dot Blot (1:10000) Western Blot (1:500-1:750) The optimal working dilution should be determined by the end user. | |
| MIG7 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction