Learn More
MIF Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MIF.
Brand: Abnova PAB7990.100ug
Description
This gene encodes a lymphokine involved in cell-mediated immunity, immunoregulation, and inflammation. It plays a role in the regulation of macrophage function in host defense through the suppression of anti-inflammatory effects of glucocorticoids. This lymphokine and the JAB1 protein form a complex in the cytosol near the peripheral plasma membrane, which may indicate an additional role in integrin signaling pathways. [provided by RefSeq
Specifications
| MIF | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MIF. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| GIF/GLIF/MMIF | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4282 | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MIF | |
| MIF | |
| A synthetic peptide corresponding to amino acids at C-terminus of human MIF. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only