missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEX3B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MEX3B.
Brand: Abnova PAB25779.100ug
This item is not returnable.
View return policy
Specifications
| MEX3B | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| MEX3B | |
| MEX3B | |
| A synthetic peptide corresponding to 16 amino acids at internal region of human MEX3B. | |
| 100 μg | |
| Primary | |
| This antibody is specific for RKHD3 and will not recognize the other RKHD3 family of proteins. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MEX3B. | |
| In PBS (0.02% sodium azide) | |
| DKFZp434J0617/MEX-3B/MGC117199/RKHD3/RNF195 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 84206 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction