missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERTK/TYRO3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MERTK/TYRO3.
Brand: Abnova PAB29209.100uL
This item is not returnable.
View return policy
Specifications
| MERTK/TYRO3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MERTK/TYRO3. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| MERTK | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y749/681 of human MERTK/TYRO3. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of MER/TYRO3 only when phosphorylated at tyrosine 749/681. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
| MER/MGC133349/RP38/c-mer | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 10461/7301 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction